Assay Details
| Gene Symbol: Usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033202.3 | NP_001028374.1 | 10 - 11 | 1051 | 72 | |
| GenBank mRNA | AK078164.1 | - | 6 - 7 | 403 | 72 | |
| AK144578.1 | - | 10 - 11 | 1052 | 72 | ||
| AK162153.1 | - | 100 | 72 | |||
| AK162269.1 | - | 3 - 4 | 221 | 72 | ||
| BC038606.1 | - | 10 - 11 | 1073 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitophagy
proteolysis
ubiquitin-dependent protein catabolic process
mitochondrial fusion
protein deubiquitination
protein K11-linked deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
protein K6-linked deubiquitination
regulation of proteasomal protein catabolic process
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
peptidase activity
cysteine-type peptidase activity
hydrolase activity
thiol-dependent ubiquitinyl hydrolase activity
mitophagy
proteolysis
ubiquitin-dependent protein catabolic process
mitochondrial fusion
protein deubiquitination
protein K11-linked deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
protein K6-linked deubiquitination
regulation of proteasomal protein catabolic process
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
peptidase activity
cysteine-type peptidase activity
hydrolase activity
thiol-dependent ubiquitinyl hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026089 | AK162153.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026089 | AK078164.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026089 | AK144578.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026089 | NM_001033202.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 65984 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270068 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816025 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135128 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838032 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 258366 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 507291 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320740 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 586861 | AK049862.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738367 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747673 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237579 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548287 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 171597 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13925 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026089 | AK162153.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026089 | AK078164.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026089 | AK144578.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026089 | NM_001033202.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01192303 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01192333 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01192339 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05702815 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01192311 | - | - |
| Whole Mouse Genome G4122A | A_52_P179272 | BC038606.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P179272 | AK162153.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P179272 | AK162269.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P179272 | AK078164.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P179272 | AK144578.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P161413 | AK162153.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P161413 | AK078164.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05702815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24002034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24001846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04297680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04703684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04297690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04224897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24002037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24002035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04530282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24002045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04297681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24002038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24002026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24002040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04703685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04297679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24002034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04297682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24002042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04297686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04297694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04930265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24002041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24001923 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10524445 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524441 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524447 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524443 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524444 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524439 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524437 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524448 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524438 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524442 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524449 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524440 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436009_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455598_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10524436 | - | - |
| Mouse Genome 430 2.0 Array | 1455598_at | - | - |
| Mouse Genome 430 2.0 Array | 1436009_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436009_at | - | - |
| Mouse Genome 430 2.0 Array | 1455598_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100756_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100756_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500010339.mm.1 | TC0500001219.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500010339.mm.1 | NM_001033202.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500010339.mm.1 | NM_001033202.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500010337.mm.1 | TC0500001219.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500010337.mm.1 | NM_001033202.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500010337.mm.1 | NM_001033202.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500010338.mm.1 | TC0500001219.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500010338.mm.1 | NM_001033202.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500010338.mm.1 | NM_001033202.3 | Refseq |
| Mouse Expression Set 430 | 1436009_PM_at | - | - |
| Mouse Expression Set 430 | 1436009_at | - | - |
| Mouse Expression Set 430 | 1455598_PM_at | - | - |
| Mouse Expression Set 430 | 1455598_at | - | - |
| Murine Genome U74v2 Set | 166233_PM_at | - | - |
| Murine Genome U74v2 Set | 166233_at | - | - |
| Murine Genome U74v2 Set | 135925_PM_at | - | - |
| Murine Genome U74v2 Set | 135925_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436009_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436009_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455598_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455598_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161250 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161250 | - | - |
| mouse-6 v1.1 (Illumina) | GI_85986574-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161250 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161250 | - | - |

Usp30
Genomic Map