Assay Details
| Gene Symbol: Usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033202.3 | NP_001028374.1 | 7 - 8 | 815 | 80 | |
| GenBank mRNA | AK078164.1 | - | 3 - 4 | 167 | 80 | |
| AK144578.1 | - | 7 - 8 | 816 | 80 | ||
| AK216289.1 | - | 3 - 4 | 181 | 80 | ||
| BC038606.1 | - | 7 - 8 | 837 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp30
Genomic Map