Assay Details
| Gene Symbol: Usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033202.3 | NP_001028374.1 | 9 - 10 | 974 | 67 | |
| GenBank mRNA | AK078164.1 | - | 5 - 6 | 326 | 67 | |
| AK144578.1 | - | 9 - 10 | 975 | 67 | ||
| AK162269.1 | - | 2 - 3 | 144 | 67 | ||
| BC038606.1 | - | 9 - 10 | 996 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp30
Genomic Map