Assay Details
| Gene Symbol: Dennd2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093754.2 | NP_001087223.1 | 7 - 8 | 1011 | 79 | |
| NM_001278941.1 | NP_001265870.1 | 8 - 9 | 1090 | 79 | ||
| NM_028110.2 | NP_082386.2 | 7 - 8 | 1009 | 79 | ||
| GenBank mRNA | AK008546.1 | - | 7 - 8 | 815 | 79 | |
| AK050033.1 | - | 8 - 9 | 1091 | 79 | ||
| AK157279.1 | - | 7 - 8 | 1012 | 79 | ||
| BC008266.1 | - | 7 - 8 | 999 | 2811014 | 79 | |
| BC043727.1 | - | 7 - 8 | 797 | 5035764 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dennd2d
Genomic Map