Assay Details
| Gene Symbol: Prok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC042707.1 | - | 1 - 2 | 45 | 4975079 | 65 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC042707.1 transcripts.
Set Membership: |
|

Prok1
Genomic Map