Assay Details
| Gene Symbol: Alx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007441.3 | NP_031467.1 | 2 - 3 | 737 | 68 | |
| GenBank mRNA | AK142019.1 | - | 2 - 3 | 663 | 68 | |
| U96109.1 | - | 2 - 3 | 740 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alx3
Genomic Map