Assay Details
| Gene Symbol: Eps8l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133867.2 | NP_598628.1 | 2 - 3 | 191 | 84 | |
| GenBank mRNA | AK136584.1 | - | 2 - 3 | 187 | 84 | |
| AK167630.1 | - | 2 - 3 | 178 | 84 | ||
| AY074932.1 | - | 2 - 3 | 207 | 84 | ||
| BC014734.1 | - | 2 - 3 | 179 | 4218079 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK167630.1, AY074932.1, BC014734.1, NM_133867.2 transcripts.
Set Membership: |
|

Eps8l3
Genomic Map