Assay Details
| Gene Symbol: Eps8l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133867.2 | NP_598628.1 | 3 - 4 | 259 | 63 | |
| GenBank mRNA | AK136584.1 | - | 3 - 4 | 255 | 63 | |
| AK167630.1 | - | 3 - 4 | 246 | 63 | ||
| AY074932.1 | - | 3 - 4 | 275 | 63 | ||
| BC014734.1 | - | 3 - 4 | 247 | 4218079 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eps8l3
Genomic Map