Assay Details
| Gene Symbol: Eps8l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133867.2 | NP_598628.1 | 6 - 7 | 625 | 61 | |
| GenBank mRNA | AK136584.1 | - | 6 - 7 | 621 | 61 | |
| AK167630.1 | - | 6 - 7 | 612 | 61 | ||
| AY074932.1 | - | 6 - 7 | 641 | 61 | ||
| BC014734.1 | - | 6 - 7 | 613 | 4218079 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eps8l3
Genomic Map