Assay Details
| Gene Symbol: Eps8l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133867.2 | NP_598628.1 | 9 - 10 | 1022 | 110 | |
| GenBank mRNA | AK136584.1 | - | 9 - 10 | 1019 | 110 | |
| AY074932.1 | - | 9 - 10 | 1038 | 110 | ||
| BC014734.1 | - | 9 - 10 | 1010 | 4218079 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eps8l3
Genomic Map