Assay Details
| Gene Symbol: Slc6a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293689.1 | NP_001280618.1 | 4 - 5 | 1077 | 86 | |
| NM_172271.2 | NP_758475.1 | 4 - 5 | 1077 | 86 | ||
| NR_121629.1 | 3 - 4 | 664 | 86 | |||
| GenBank mRNA | AK032262.1 | - | 3 - 4 | 454 | 86 | |
| AK083807.1 | - | 4 - 5 | 978 | 86 | ||
| AK085620.1 | - | 4 - 5 | 811 | 86 | ||
| AK163113.1 | - | 4 - 5 | 1082 | 86 | ||
| AY155578.1 | - | 4 - 5 | 1078 | 86 | ||
| BC033610.1 | - | 4 - 5 | 702 | 86 | ||
| BC141148.1 | - | 3 - 4 | 648 | 9055760 | 86 | |
| BC171962.1 | - | 3 - 4 | 648 | 9054248 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a17
Genomic Map