Assay Details
| Gene Symbol: Cym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111143.1 | NP_001104613.1 | 1 - 2 | 90 | 62 | |
| GenBank mRNA | BC147477.1 | - | 1 - 2 | 87 | 9056130 | 62 |
| BC171994.1 | - | 1 - 2 | 87 | 9054280 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cym
Genomic Map