Assay Details
| Gene Symbol: Cym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111143.1 | NP_001104613.1 | 7 - 8 | 933 | 70 | |
| GenBank mRNA | BC147477.1 | - | 7 - 8 | 930 | 9056130 | 70 |
| BC171994.1 | - | 7 - 8 | 930 | 9054280 | 70 | |
More Information
Set Membership: |
|

Cym
Genomic Map