Assay Details
| Gene Symbol: Cym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111143.1 | NP_001104613.1 | 8 - 9 | 1035 | 66 | |
| GenBank mRNA | BC147477.1 | - | 8 - 9 | 1032 | 9056130 | 66 |
| BC171994.1 | - | 8 - 9 | 1032 | 9054280 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046563 | NM_001111143.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046563 | BC147477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046563 | BC171994.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122429 | NM_001111143.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910196 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 428077 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 842168 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28598 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140851 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 689468 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369290 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119347 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046563 | NM_001111143.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046563 | BC147477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046563 | BC171994.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122429 | NM_001111143.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P395492 | NM_001111143.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P395492 | BC147477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P395492 | BC171994.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22404926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03094811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02339818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02529683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02604936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03199476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02529678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02322920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03140746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02653925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03063336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02653067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02611106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03001580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02875642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03063338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02878226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02611101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02604940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02653066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02878225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03063339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03001573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03199477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03094812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02641304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02530647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22405040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22404965 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10501082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501080 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501079 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501083 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501076 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501078 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501081 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501077 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501075 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457271_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501074 | - | - |
| Mouse Genome 430 2.0 Array | 1457271_at | - | - |
| Mouse Genome 430 2.0 Array | 1457271_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229697_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229697_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300021028.mm.1 | NM_001111143.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021028.mm.1 | NM_001111143.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021028.mm.1 | TC0300002701.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021030.mm.1 | NM_001111143.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021030.mm.1 | NM_001111143.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021030.mm.1 | TC0300002701.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021029.mm.1 | NM_001111143.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021029.mm.1 | NM_001111143.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021029.mm.1 | TC0300002701.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1457271_at | - | - |
| Mouse Expression Set 430 | 1457271_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 735662 | - | - |

Cym
Genomic Map