Assay Details
| Gene Symbol: Strip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153563.2 | NP_705791.2 | 2 - 3 | 277 | 71 | |
| GenBank mRNA | AK032065.1 | - | 2 - 3 | 259 | 71 | |
| AK035833.1 | - | 2 - 3 | 262 | 71 | ||
| AK122553.1 | - | 2 - 3 | 241 | 71 | ||
| AK148011.1 | - | 2 - 3 | 262 | 71 | ||
| AK169644.1 | - | 2 - 3 | 266 | 71 | ||
| AK191730.1 | - | 2 - 3 | 91 | 71 | ||
| AK194900.1 | - | 2 - 3 | 131 | 71 | ||
| AK200873.1 | - | 1 - 2 | 91 | 71 | ||
| AK213419.1 | - | 2 - 3 | 91 | 71 | ||
| BC023952.1 | - | 2 - 3 | 253 | 5320305 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC023952.1 transcripts.
Set Membership: |
|

Strip1
Genomic Map