Assay Details
| Gene Symbol: Strip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153563.2 | NP_705791.2 | 6 - 7 | 681 | 83 | |
| GenBank mRNA | AK032065.1 | - | 6 - 7 | 663 | 83 | |
| AK035833.1 | - | 6 - 7 | 666 | 83 | ||
| AK122553.1 | - | 6 - 7 | 645 | 83 | ||
| AK148011.1 | - | 6 - 7 | 666 | 83 | ||
| AK169644.1 | - | 6 - 7 | 613 | 83 | ||
| AK178114.1 | - | 1 - 2 | 61 | 83 | ||
| BC023952.1 | - | 6 - 7 | 657 | 5320305 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Strip1
Genomic Map