Assay Details
| Gene Symbol: Tusc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030254.3 | NP_084530.3 | 998 | 63 | ||
| GenBank mRNA | AK028182.1 | - | 974 | 63 | ||
| AK088616.1 | - | 1012 | 63 | |||
| AK132766.1 | - | 1021 | 63 | |||
| BC069898.1 | - | 998 | 6489807 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tusc3
Genomic Map