Assay Details
| Gene Symbol: Rorb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289921.1 | NP_001276850.1 | 1 - 2 | 114 | 59 | |
| GenBank mRNA | BC024768.1 | - | 1 - 2 | 97 | 59 | |
| BC024842.1 | - | 1 - 2 | 114 | 5358124 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC024842.1, NM_001289921.1 transcripts.
Set Membership: |
|

Rorb
Genomic Map