Assay Details
| Gene Symbol: Cyp11b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033229.3 | NP_001028401.2 | 3 - 4 | 630 | 69 | |
| GenBank mRNA | AK142439.1 | - | 3 - 4 | 607 | 69 | |
| AK163083.1 | - | 3 - 4 | 606 | 69 | ||
| AK168839.1 | - | 3 - 4 | 607 | 69 | ||
| BC139248.1 | - | 3 - 4 | 630 | 8862270 | 69 | |
| BC139250.1 | - | 3 - 4 | 630 | 8862272 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp11b1
Genomic Map