Assay Details
| Gene Symbol: Fas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007987.2 | NP_032013.2 | 3 - 4 | 372 | 101 | |
| GenBank mRNA | AK002590.1 | - | 3 - 4 | 323 | 101 | |
| BC061160.1 | - | 3 - 4 | 345 | 30302649 | 101 | |
| DQ846748.1 | - | 3 - 4 | 316 | 101 | ||
| M83649.1 | - | 3 - 4 | 365 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fas
Genomic Map