Assay Details
| Gene Symbol: Lipa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111100.1 | NP_001104570.1 | 3 - 4 | 334 | 59 | |
| NM_021460.3 | NP_067435.3 | 3 - 4 | 338 | 59 | ||
| GenBank mRNA | AK088659.1 | - | 3 - 4 | 290 | 59 | |
| AK149791.1 | - | 3 - 4 | 298 | 59 | ||
| AK154804.1 | - | 3 - 4 | 280 | 59 | ||
| AK164007.1 | - | 3 - 4 | 334 | 59 | ||
| AK169567.1 | - | 3 - 4 | 304 | 59 | ||
| BC058564.1 | - | 3 - 4 | 286 | 6441278 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lipa
Genomic Map