Assay Details
| Gene Symbol: As3mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020577.2 | NP_065602.2 | 1 - 2 | 98 | 70 | |
| GenBank mRNA | AF166383.1 | - | 1 - 2 | 68 | 70 | |
| AK006571.1 | - | 1 - 2 | 83 | 70 | ||
| AK011320.1 | - | 1 - 2 | 50 | 70 | ||
| BC145663.1 | - | 1 - 2 | 40 | 40130966 | 70 | |
| BY287843.1 | - | 1 - 2 | 98 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF166383.1, BC145663.1, NM_020577.2 transcripts.
Set Membership: |
|

As3mt
Genomic Map