Assay Details
| Gene Symbol: As3mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020577.2 | NP_065602.2 | 8 - 9 | 842 | 86 | |
| GenBank mRNA | AF166383.1 | - | 8 - 9 | 812 | 86 | |
| AK002801.1 | - | 8 - 9 | 84 | |||
| AK006571.1 | - | 8 - 9 | 827 | 86 | ||
| AK009814.1 | - | 7 - 8 | 807 | 86 | ||
| BC013468.1 | - | 8 - 9 | 767 | 86 | ||
| BC145663.1 | - | 8 - 9 | 784 | 40130966 | 86 | |
| BQ445127.1 | - | 1 - 2 | 60 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

As3mt
Genomic Map