Assay Details
| Gene Symbol: Smurf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038627.1 | NP_001033716.1 | 5 - 6 | 671 | 95 | |
| NM_029438.3 | NP_083714.3 | 5 - 6 | 671 | 95 | ||
| GenBank mRNA | AK154491.1 | - | 5 - 6 | 672 | 95 | |
| BC029097.1 | - | 5 - 6 | 699 | 3660965 | 95 | |
| CX568994.1 | - | 3 - 4 | 271 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smurf1
Genomic Map