Assay Details
| Gene Symbol: Smurf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038627.1 | NP_001033716.1 | 7 - 8 | 997 | 81 | |
| NM_029438.3 | NP_083714.3 | 7 - 8 | 997 | 81 | ||
| GenBank mRNA | AK154491.1 | - | 7 - 8 | 998 | 81 | |
| BC029097.1 | - | 7 - 8 | 1025 | 3660965 | 81 | |
| CB524571.1 | - | 1 - 2 | 75 | 81 | ||
| CX568994.1 | - | 5 - 6 | 597 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smurf1
Genomic Map