Assay Details
| Gene Symbol: Smurf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038627.1 | NP_001033716.1 | 9 - 10 | 1227 | 64 | |
| NM_029438.3 | NP_083714.3 | 9 - 10 | 1227 | 64 | ||
| GenBank mRNA | AK015264.1 | - | 5 - 6 | 626 | 64 | |
| AK154491.1 | - | 9 - 10 | 1228 | 64 | ||
| BC029097.1 | - | 9 - 10 | 1255 | 3660965 | 64 | |
| CB524571.1 | - | 3 - 4 | 305 | 64 | ||
| CB527254.1 | - | 1 - 2 | 151 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smurf1
Genomic Map