Assay Details
| Gene Symbol: Ddx51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027156.3 | NP_081432.2 | 11 - 12 | 1627 | 84 | |
| GenBank mRNA | AK010015.1 | - | 2 - 3 | 231 | 84 | |
| AK141361.1 | - | 11 - 12 | 1608 | 84 | ||
| AK152599.1 | - | 11 - 12 | 1595 | 84 | ||
| BC023700.1 | - | 8 - 9 | 890 | 84 | ||
| BC060646.1 | - | 11 - 12 | 1604 | 6853826 | 84 | |
| BC069876.1 | - | 4 - 5 | 455 | 84 | ||
More Information
Set Membership: |
|

Ddx51
Genomic Map