Assay Details
| Gene Symbol: Ddx51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027156.3 | NP_081432.2 | 6 - 7 | 946 | 115 | |
| GenBank mRNA | AK141361.1 | - | 6 - 7 | 927 | 115 | |
| AK152599.1 | - | 6 - 7 | 914 | 115 | ||
| BC023700.1 | - | 3 - 4 | 209 | 115 | ||
| BC060646.1 | - | 6 - 7 | 923 | 6853826 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx51
Genomic Map