Assay Details
| Gene Symbol: Ulk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009469.3 | NP_033495.2 | 4 - 5 | 632 | 76 | |
| GenBank mRNA | AF053756.1 | - | 4 - 5 | 324 | 76 | |
| AF072370.1 | - | 4 - 5 | 315 | 76 | ||
| AK122357.1 | - | 3 - 4 | 134 | 76 | ||
| AK148317.1 | - | 4 - 5 | 607 | 76 | ||
| AK154939.1 | - | 4 - 5 | 649 | 76 | ||
| AK156534.1 | - | 4 - 5 | 609 | 76 | ||
| BC053062.1 | - | 4 - 5 | 292 | 76 | ||
| BC057121.1 | - | 4 - 5 | 632 | 6834534 | 76 | |
| BC059835.1 | - | 4 - 5 | 583 | 6406755 | 76 | |
More Information
Set Membership: |
|

Ulk1
Genomic Map