Assay Details
| Gene Symbol: Hscb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153571.2 | NP_705799.2 | 5 - 6 | 657 | 97 | |
| GenBank mRNA | AK131961.1 | - | 5 - 6 | 659 | 97 | |
| BC027641.1 | - | 5 - 6 | 621 | 1244839 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23982205 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23982207 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105220 | AK131961.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105220 | BC027641.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105220 | NM_153571.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 373742 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 165289 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 757657 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86020 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83179 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 122791 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 20309 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134070 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167185 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105220 | AK131961.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105220 | BC027641.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105220 | NM_153571.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P298052 | AK131961.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P298052 | BC027641.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P298052 | NM_153571.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23982203 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167185 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134070 | - | - |
| Whole Mouse Genome G4122A | A_51_P298052 | AK131961.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P298052 | BC027641.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P298052 | NM_153571.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140661 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139401 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167185 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134070 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04934410 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04934411 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04665779 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04665776 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23982205 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23982207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23982174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04782889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04601718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04337326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04782888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04782885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04934411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04688889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04509304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04962838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04504065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23982195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04665779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04337319 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04337323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04322029 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10532510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532508 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532506 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532505 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532507 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427927_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10532504 | - | - |
| Mouse Genome 430 2.0 Array | 1427927_at | - | - |
| Mouse Genome 430 2.0 Array | 1427927_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100900_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100900_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500026641.mm.1 | TC0500003001.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026641.mm.1 | NM_153571.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026641.mm.1 | NM_153571.2 | Refseq |
| Mouse Expression Set 430 | 1427927_at | - | - |
| Mouse Expression Set 430 | 1427927_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427927_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427927_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427927_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427927_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 555617 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1400750 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1400750 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4210162 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004210162 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001400750 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2611037 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2740652 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2740652 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2611037 | - | - |
| mouse-6 v1.1 (Illumina) | scl0004041.1_32-S | - | - |
| mouse-6 v1.1 (Illumina) | scl26218.7.1_61-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4210162 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1400750 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26218.7.1_61-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0004041.1_32-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4210162 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1400750 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2611037 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2740652 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2740652 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2611037 | - | - |

Hscb
Genomic Map