Assay Details
| Gene Symbol: Trim28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011588.3 | NP_035718.2 | 13 - 14 | 2545 | 81 | |
| GenBank mRNA | AF230391.1 | - | 13 - 14 | 2146 | 81 | |
| AK089084.1 | - | 1 - 2 | 197 | 81 | ||
| AK141357.1 | - | 13 - 14 | 2289 | 81 | ||
| AK149995.1 | - | 10 - 11 | 1370 | 81 | ||
| AK181992.1 | - | 1 - 2 | 284 | 81 | ||
| BC020452.1 | - | 13 - 14 | 1976 | 81 | ||
| BC023335.1 | - | 13 - 14 | 2254 | 81 | ||
| BC058391.1 | - | 13 - 14 | 1991 | 6817165 | 81 | |
| BC089337.1 | - | 13 - 14 | 2545 | 6811294 | 81 | |
| U67303.1 | - | 13 - 14 | 2536 | 81 | ||
| X99644.1 | - | 13 - 14 | 2146 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim28
Genomic Map