Assay Details
| Gene Symbol: Trim28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011588.3 | NP_035718.2 | 2 - 3 | 1026 | 85 | |
| GenBank mRNA | AF230391.1 | - | 2 - 3 | 627 | 85 | |
| AF230392.1 | - | 2 - 3 | 627 | 85 | ||
| AK141357.1 | - | 2 - 3 | 770 | 85 | ||
| AK184928.1 | - | 1 - 2 | 131 | 85 | ||
| AK217247.1 | - | 2 - 3 | 131 | 85 | ||
| AK219334.1 | - | 1 - 2 | 131 | 85 | ||
| BC020452.1 | - | 2 - 3 | 457 | 85 | ||
| BC023335.1 | - | 2 - 3 | 735 | 85 | ||
| BC058391.1 | - | 2 - 3 | 472 | 6817165 | 85 | |
| BC089337.1 | - | 2 - 3 | 1026 | 6811294 | 85 | |
| U67303.1 | - | 2 - 3 | 1017 | 85 | ||
| X99644.1 | - | 2 - 3 | 627 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim28
Genomic Map