Assay Details
| Gene Symbol: Trim28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011588.3 | NP_035718.2 | 7 - 8 | 1672 | 80 | |
| GenBank mRNA | AF230391.1 | - | 7 - 8 | 1273 | 80 | |
| AF230392.1 | - | 7 - 8 | 1273 | 80 | ||
| AK141357.1 | - | 7 - 8 | 1416 | 80 | ||
| AK149995.1 | - | 4 - 5 | 497 | 80 | ||
| BC020452.1 | - | 7 - 8 | 1103 | 80 | ||
| BC023335.1 | - | 7 - 8 | 1381 | 80 | ||
| BC058391.1 | - | 7 - 8 | 1118 | 6817165 | 80 | |
| BC089337.1 | - | 7 - 8 | 1672 | 6811294 | 80 | |
| U67303.1 | - | 7 - 8 | 1663 | 80 | ||
| X99644.1 | - | 7 - 8 | 1273 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim28
Genomic Map