Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700006930.mm.1 |
TC0700000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006930.mm.1 |
NM_027376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006930.mm.1 |
NM_027376.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006921.mm.1 |
TC0700000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006921.mm.1 |
NM_027376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006921.mm.1 |
NM_027376.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006923.mm.1 |
TC0700000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006923.mm.1 |
NM_027376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006923.mm.1 |
NM_027376.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006920.mm.1 |
TC0700000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006920.mm.1 |
NM_027376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006920.mm.1 |
NM_027376.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006926.mm.1 |
TC0700000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006926.mm.1 |
NM_027376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006926.mm.1 |
NM_027376.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006922.mm.1 |
TC0700000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006922.mm.1 |
NM_027376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006922.mm.1 |
NM_027376.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006928.mm.1 |
TC0700000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006928.mm.1 |
NM_027376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006928.mm.1 |
NM_027376.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006927.mm.1 |
TC0700000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006927.mm.1 |
NM_027376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006927.mm.1 |
NM_027376.3 |
Refseq |