Assay Details
| Gene Symbol: Fuz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027376.3 | NP_081652.2 | 9 - 10 | 1133 | 94 | |
| GenBank mRNA | AK033247.1 | - | 9 - 10 | 1120 | 94 | |
| AK045745.1 | - | 1128 | 94 | |||
| CK624547.1 | - | 4 - 5 | 395 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fuz
Genomic Map