Assay Details
| Gene Symbol: Vrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133945.1 | NP_598706.1 | 10 - 11 | 985 | 76 | |
| GenBank mRNA | AK150435.1 | - | 10 - 11 | 1076 | 76 | |
| AK171673.1 | - | 10 - 11 | 990 | 76 | ||
| AK183463.1 | - | 3 - 4 | 253 | 76 | ||
| BC010473.1 | - | 9 - 10 | 968 | 3586304 | 76 | |
| BC024782.1 | - | 9 - 10 | 968 | 5123949 | 76 | |
| BC024839.1 | - | 10 - 11 | 985 | 5355521 | 76 | |
| BC027129.1 | - | 9 - 10 | 914 | 76 | ||
| BC034196.1 | - | 9 - 10 | 966 | 4985176 | 76 | |
| S71575.1 | - | 2 - 3 | 205 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vrk3
Genomic Map