Assay Details
| Gene Symbol: Pnkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290767.1 | NP_001277696.1 | 3 - 4 | 494 | 99 | |
| GenBank mRNA | AF129451.1 | - | 3 - 4 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290767.1 transcripts.
Set Membership: |
|

Pnkp
Genomic Map