Assay Details
| Gene Symbol: Slc17a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182993.2 | NP_892038.2 | 3 - 4 | 596 | 99 | |
| GenBank mRNA | AK159064.1 | - | 3 - 4 | 569 | 99 | |
| BC054462.1 | - | 1 - 2 | 378 | 6494471 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC054462.1 transcripts.
Set Membership: |
|

Slc17a7
Genomic Map