Assay Details
| Gene Symbol: Ruvbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011304.3 | NP_035434.1 | 6 - 7 | 667 | 76 | |
| GenBank mRNA | AB013912.1 | - | 6 - 7 | 498 | 76 | |
| AK135404.1 | - | 6 - 7 | 481 | 76 | ||
| AK144156.1 | - | 6 - 7 | 489 | 76 | ||
| AK159112.1 | - | 6 - 7 | 669 | 76 | ||
| BC072585.1 | - | 1166 | 76 | |||
| BC115809.1 | - | 6 - 7 | 485 | 40105495 | 76 | |
| BC116693.1 | - | 6 - 7 | 484 | 40105493 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ruvbl2
Genomic Map