Assay Details
| Gene Symbol: Izumo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018013.1 | NP_001018013.1 | 1 - 2 | 187 | 81 | |
| GenBank mRNA | AB195681.1 | - | 1 - 2 | 187 | 81 | |
| AK006830.1 | - | 1 - 2 | 187 | 81 | ||
| BC139270.1 | - | 1 - 2 | 71 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB195681.1, AK006830.1, NM_001018013.1 transcripts.
Set Membership: |
|

Izumo1
Genomic Map