Assay Details
| Gene Symbol: Izumo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018013.1 | NP_001018013.1 | 5 - 6 | 683 | 118 | |
| GenBank mRNA | AB195681.1 | - | 5 - 6 | 683 | 118 | |
| AK006830.1 | - | 5 - 6 | 683 | 118 | ||
| BC139270.1 | - | 5 - 6 | 567 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Izumo1
Genomic Map