Assay Details
| Gene Symbol: Izumo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018013.1 | NP_001018013.1 | 9 - 10 | 1277 | 66 | |
| GenBank mRNA | AB195681.1 | - | 9 - 10 | 1277 | 66 | |
| AK006830.1 | - | 9 - 10 | 1277 | 66 | ||
| BC139270.1 | - | 8 - 9 | 1480 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098423 | AB195681.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098423 | BC139270.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098423 | NM_001018013.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61359 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331283 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697040 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 222394 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23211 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137504 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098423 | AB195681.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098423 | BC139270.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098423 | NM_001018013.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094106 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137504 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01508054 | - | - |
| Whole Mouse Genome G4122A | A_51_P486350 | AB195681.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486350 | AK006830.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486350 | BC139270.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486350 | NM_001018013.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126339 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137504 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06633543 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06633544 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06156580 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06156588 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06633550 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06156584 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06633548 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06156581 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06497471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06156581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06094508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06633548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06497471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06094521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06633545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06156588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094089 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10553040 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553036 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553039 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553041 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553034 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553032 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553033 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553035 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553038 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429695_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10553031 | - | - |
| Mouse Genome 430 2.0 Array | 1429695_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429695_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73456_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73456_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700007474.mm.1 | TC0700000856.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700007474.mm.1 | NM_001018013.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007474.mm.1 | NM_001018013.1 | Refseq |
| Mouse Expression Set 430 | 1429695_at | - | - |
| Mouse Expression Set 430 | 1429695_PM_at | - | - |
| Murine Genome U74v2 Set | 170164_r_at | - | - |
| Murine Genome U74v2 Set | 171336_PM_i_at | - | - |
| Murine Genome U74v2 Set | 170164_PM_r_at | - | - |
| Murine Genome U74v2 Set | 171336_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429695_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429695_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 454052 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2881054 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2881054 | - | - |

Izumo1
Genomic Map