Assay Details
| Gene Symbol: Mamstr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172418.2 | NP_766006.2 | 1 - 2 | 58 | 69 | |
| GenBank mRNA | AK017382.1 | - | 1 - 2 | 59 | 69 | |
| BY717195.1 | - | 1 - 2 | 59 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_172418.2 transcripts.
Set Membership: |
|

Mamstr
Genomic Map