Assay Details
| Gene Symbol: Mamstr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172418.2 | NP_766006.2 | 2 - 3 | 198 | 67 | |
| GenBank mRNA | DQ534901.1 | - | 2 - 3 | 182 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mamstr
Genomic Map