Assay Details
| Gene Symbol: Kdelr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025841.4 | NP_080117.1 | 1 - 2 | 235 | 69 | |
| GenBank mRNA | AJ278133.1 | - | 1 - 2 | 228 | 69 | |
| AK003515.1 | - | 1 - 2 | 230 | 69 | ||
| AK007403.1 | - | 1 - 2 | 219 | 69 | ||
| AK150402.1 | - | 1 - 2 | 230 | 69 | ||
| BC007146.1 | - | 1 - 2 | 217 | 3496891 | 69 | |
| CJ173962.1 | - | 1 - 2 | 280 | 69 | ||
| CT010232.1 | - | 1 - 2 | 93 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kdelr2
Genomic Map