Assay Details
| Gene Symbol: Mmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175217.6 | NP_780426.1 | 1 - 2 | 200 | 102 | |
| GenBank mRNA | AK132153.1 | - | 249 | 102 | ||
| AY134664.1 | - | 1 - 2 | 141 | 102 | ||
| AY424299.1 | - | 1 - 2 | 200 | 102 | ||
| BC025064.1 | - | 1 - 2 | 200 | 4502289 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK132153.1, AY134664.1, AY424299.1, BC025064.1, NM_175217.6 transcripts.
Set Membership: |
|

Mmd2
Genomic Map