Assay Details
| Gene Symbol: Mmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175217.6 | NP_780426.1 | 3 - 4 | 435 | 67 | |
| GenBank mRNA | AK132153.1 | - | 484 | 67 | ||
| AY134664.1 | - | 3 - 4 | 376 | 67 | ||
| AY424299.1 | - | 3 - 4 | 435 | 67 | ||
| BC025064.1 | - | 3 - 4 | 435 | 4502289 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mmd2
Genomic Map