Assay Details
| Gene Symbol: Kifc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010630.2 | NP_034760.2 | 11 - 12 | 1242 | 85 | |
| GenBank mRNA | AK144304.1 | - | 12 - 13 | 1401 | 85 | |
| D49545.1 | - | 11 - 12 | 1318 | 85 | ||
| U92949.1 | - | 11 - 12 | 1232 | 85 | ||
More Information
Set Membership: |
|

Kifc2
Genomic Map