Assay Details
| Gene Symbol: Kifc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010630.2 | NP_034760.2 | 13 - 14 | 1518 | 68 | |
| GenBank mRNA | AK144304.1 | - | 14 - 15 | 1677 | 68 | |
| D49545.1 | - | 13 - 14 | 1594 | 68 | ||
| U92949.1 | - | 13 - 14 | 1508 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kifc2
Genomic Map