Assay Details
| Gene Symbol: Kifc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010630.2 | NP_034760.2 | 3 - 4 | 358 | 115 | |
| GenBank mRNA | AK144304.1 | - | 4 - 5 | 517 | 115 | |
| D49545.1 | - | 3 - 4 | 434 | 115 | ||
| U92949.1 | - | 3 - 4 | 348 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kifc2
Genomic Map